Learn More
SHOWA 4561 Nitrile Knit Gloves
Product Code. 17831171
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
The SHOWA 4561 cut protection glove is built to withstand harsh conditions with its Dupont™ Kevlar™ cut-resistant liner and sponge nitrile palm coating. It provides excellent cut, abrasion, and flame resistance while maintaining comfort and precision.
  • The sponge nitrile palm coating provides a strong grip, even in wet or greasy environments.
  • The seamless knit reduces irritation, and its elasticated, form-fitting design keeps dirt out.
Specifications
Construction Seamless Knit
Coating Material Nitrile
Color Yellow, Black
Hand Specific
Latex Free Yes
Material Nitrile, Kevlar
Product Type Cut-resistant Gloves
Size Medium
Gender Unisex
Glove Cuff Style Knit