missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Shimadzu GC-2010 Liners

Provides excellent analysis for a wide range of compounds.

Brand:  Trajan Scientific and Medical™ 0920425

 View more versions of this product

Product Code. 15219013

  • 376.00€ / Pack of 5
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Shimadzu instruments
Specifications

Specifications

GC-2010 Liners
1.5 mm
3 Baffle PTV for Shimadzu
Inlet liners PTV 3 baffle
3.1 mm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions