missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Shelves for Refrigerator

Product Code. 12597096 Shop All Liebherr Products
Click to view available options
Type:
Glass Shelf
Stainless Steel Shelf
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12597096

Brand: Liebherr™ 7112415

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Type Stainless Steel Shelf
Cabinet Material Stainless Steel
Color White
For Use With (Equipment) Model FKvsl 5413
Width (Metric) Exterior 645 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.