Learn More
Fisherbrand™ Isotemp™ Shelf
Material: Wire; For Use With: Isotemp gravity convection oven/Incubator, 3.7/4.0 cu. ft.
Brand: Fisherbrand™ 50145850
133.84 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Material: Stainless steel
Specifications
Isotemp gravity convection oven/incubator, 3.7/4.0 cu. ft. | |
Wire | |
Shelf | |
16.61 x 15.20 in. (422 x 386 mm) |
Promotions
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Product Title
Fisherbrand™ Isotemp™ Shelf > Wire mesh shelf for 3.7/4.0 cu. ft. gravity convection oven/incubator