Learn More
Sharpsguard™ cyto 11.5 Multi-Purpose 9.27L Waste Container
Product Code. 12342449
Click to view available options
Color:
Purple Closure
Unit Size:
Each
Product Code. 12342449
Brand: Daniels Healthcare DD610
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
An ideal multi-purpose container for cytotoxic and cytostatic waste. Sharpsguard™ cyto 11.5 Multi-Purpose 9.27L Waste Container has a robust pail handle for easy transportation.
  • Supplied with an absorbent pad
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection against penetrating sharps and leaking fluids
  • When container is full to the fill line it must be locked and disposed of safely
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications
Type Waste Bin
Material Polypropylene
Color Purple Closure
Shape Round
Capacity (Metric) 11.5 (Overall) L
Closure Type Click Lid
Includes Absorbent Pad, Click Lid, Handle