missing translation for 'onlineSavingsMsg'
Learn More

Hospidex™ Sharpsafe™ Polypropylene Biohazard Container

Product Code. 11972089
Click to view available options
Capacity (Metric):
1 L
2 L
7 L
13 L
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 50
This item is not returnable. View return policy

Product Code. 11972089

Brand: Hospidex™ SHA4160

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Use the Sharpsafe™ Polypropylene Biohazard Container to transport dangerous biohazardous goods for disposal.

  • Yellow
  • Polypropylene
  • NFX 30-505 certification

Specifications

Capacity (Metric) 1 L
Certifications/Compliance NFX30-500
Height (Metric) 188 mm
Length (Metric) 106 mm
Autoclavable Autoclavable
For Use With (Application) For collection of sharps waste treatment activities, pungent, sharp and / or pointed
Material Polypropylene
Product Type Biohazard Container
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.