Learn More
Hospidex™ Sharpsafe™ Polypropylene Biohazard Container
Product Code. 11905386
Click to view available options
Capacity (Metric):
1 L
2 L
7 L
13 L
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 50
Product Code. 11905386
Brand: Hospidex™ SHA4115A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use the Sharpsafe™ Polypropylene Biohazard Container to transport dangerous biohazardous goods for disposal.
  • Yellow
  • Polypropylene
  • NFX 30-505 certification
Specifications
Capacity (Metric) 13 L
Certifications/Compliance NFX30-500
Height (Metric) 294 mm
Length (Metric) 355 mm
Autoclavable Autoclavable
For Use With (Application) For collection of sharps waste treatment activities, pungent, sharp and / or pointed
Material Polypropylene
Product Type Biohazard Container