Learn More
Micronova™ Sharpie™ UltraFine Point Lab Markers
Product Code. 17685915 Shop All Micronova Products
Click to view available options
Color:
Black
Blue
Unit Size:
Pack of 12
Product Code. 17685915
Brand: Micronova™ undefined
Only null left
Add to basket
Add to basket
Sharpie™ UltraFine Point Lab Markers use permanent ink and are supplied capped. The ultra fine tip allows for precise labeling and documentation.
  • Product Type: Marker Pen
  • Tip Type: Fiber
  • Tip Size: Ultra Fine (0.2 mm)
  • Supplied with Cap: Yes
  • For Use With (Application): Labeling and Documentation
  • For Use With (Equipment): Sample Containers, Slides, Glass, Plastic, and Paper
Specifications
Material Plastic
Color Blue
Cover Color White
Dimensions (L x W) 15.24 x 1.27 cm (6 x 0.5 in.)
Autoclavable Not Autoclavable
Certifications/Compliance ISO9001 Certified Facility
Disposable Reusable
For Use With (Application) Labeling and Documentation
For Use With (Equipment) Sample Containers, Slides, Glass, Plastic, and Paper
Packaging 4 Pens/First Bag, 12 Pens/Double Bag, 12 Pens/Cs.
Product Type Permanent Marker Pen
Sterility Non-sterile
Tip Style Ultra Fine (0.2 mm)
Weight (English) 0.32 oz.
Weight (Metric) 9.0718 g