Learn More
Epredia™ Shandon™ Gill™ Hematoxylin
Achieve fast and convenient results, while maintaining complete control over stain intensity with Epredia™ Shandon™ Gill™ Hematoxylin, which has mucoid staining.
Brand: Epredia™ 6765006
95.20 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
Description
Gill 1: Light blue nuclei
Gill 2: Medium blue nuclei
Gill 3: Blue-black nuclei
Specifications
Shandon Gill™ 1 Hematoxylin | |
4L |
Safety and Handling
- Shandon Gill Hematoxylin 1
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.