Learn More
Epredia™ Shandon™ Disposable Base Molds, 0.95 x 0.95 x 0.2 in. (24 x 24 x 5mm)
Product Code. 11356810 Shop All Epredia Products
Click to view available options
Length (Metric):
7 mm
15 mm
24 mm
Width (Metric):
7 mm
15 mm
24 mm
30 mm
37 mm
Unit Size:
Pack of 500
Product Code. 11356810
Brand: Epredia™ 41744
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Promote rapid paraffin hardening with Epredia™ Shandon™ Disposable Base Molds.
  • Constructed of rigid transparent plastic
TRUSTED_SUSTAINABILITY
Specifications
Product Type Base Mold
Material Plastic
Length (Metric) 24 mm
Width (Metric) 37 mm
Length (English) 0.95 in.
Width (English) 1.46 in.
Depth (Metric) 5 mm
Depth (English) 0.2 in.
Disposable Single-use