Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs, aqua
Product Code. 12963765 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Fluorescent Green
Fluorescent Pink
Fluorescent Yellow
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Unit Size:
Each
Pack of 750
Product Code. 12963765
Brand: Epredia™ B1000730FPK
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Aid the process of speedy and reliable biopsy specimen labeling with Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs.
  • Writing face is flat 45° angle
  • Locking lid attached by two hinges
  • Front or back opening
Ordering Information:
  • Packaged in 75-cassette tubes
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Square Pore
Writing Face Angle 45°
Color Fluorescent Pink
Product Type Biopsy Cassette