Learn More
Novus Biologicals™ SGPL1 Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-07936
Product Code. 18079788
  • 685.73€ / 0.10mg
Explore more special offers
Description
Description
The SGPL1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_3892. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens SGPL1, mRNA.
Native