Learn More
Novus Biologicals™ SEZ6/BSRP-C Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-04939
Product Code. 18076167
  • 766.72€ / 0.10mg
Explore more special offers
Description
Description
The SEZ6/BSRP-C Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_849191. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens SEZ6, mRNA.
Native