missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17807-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene interacts with target proteins during their translocation into the lumen of the endoplasmic reticulum. Protects unfolded target proteins against degradation during ER stress. May facilitate glycosylation of target proteins after termination of ER stress. May modulate the use of N-glycosylation sites on target proteins.Specifications
| SERP1 | |
| Polyclonal | |
| Unconjugated | |
| SERP1 | |
| RAMP4, SERP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27230, 28146, 80881 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9R2C1, Q9Y6X1, Q9Z1W5 | |
| Serp1 | |
| SERP1 Fusion Protein Ag12087 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction