Learn More
Fisherbrand™ Septum for 10-425 screw thread cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand: Fisherbrand™ 09 02 1260
Description
- Septa
Specifications
Septa | |
9.4 mm | |
1000 |
Red, White | |
PTFE, Silicone | |
1.3 mm |