Learn More
Applied Biosystems™ Septa for 3500 Dx/3500xL Dx Genetic Analyzers, 384 well
This is a 384-well septa designed for use with the 3500 Dx/3500xL Dx Genetic Analyzer
Brand:  Applied Biosystems™ 4410607
Product Code. 17226493
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • This is a 384-well septa designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
384-Well Microtiter Plates, MicroAmp™ Optical 384-Well Reaction Plates
384-well
Septa
Room Temperature
Ambient storage.
Each package contains 20 Septas.
3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
20 Pk.