Learn More
Applied Biosystems™ Septa for 3500 Dx/3500xL Dx Genetic Analyzers, 384 well
This is a 384-well septa designed for use with the 3500 Dx/3500xL Dx Genetic Analyzer
Brand: Applied Biosystems™ 4410607
This item is not returnable.
View return policy
Description
- This is a 384-well septa designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer
- Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers
- The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
Specifications
384-Well Microtiter Plates, MicroAmp™ Optical 384-Well Reaction Plates | |
384-well | |
Septa | |
Room Temperature |
Ambient storage. Each package contains 20 Septas. |
|
3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer | |
20 Pk. |