Learn More
BD Sentry™ Safety Lancet
Allows for quick and safe blood collection from the fingertip
Brand:  BD 369523
82.94 EUR valid until 2025-05-18
Use promo code "24096" to get your promotional price.
Product Code. 16611592
  • 219.00€ / Pack of 100
Edge
Explore more special offers
Description
Description
  • Activated by the push of a button
  • Mechanism to prevent reuse and needlestick injuries
  • Easy disposal in sharps containers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Green
Single-use
100/Pk.
1.8 mm
23G
Sterile
Promotions
  • Multiple Manufacturers
    Use code 24096 to receive this promotion.
    Promo Code: 24096
    Expires2025-05-16
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 21/04/2025 and 16/05/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions Learn More
Product Content Correction
Product Title
BD Sentry™ Safety Lancet > Sterile, 23G, 1.8 mm, Green, 100/Pk.