Learn More
WTW™ SenTix™ Epoxy Combination pH Electrode
For use with WTW ProfiLine and inoLab™ pH meter series, this SenTix™ Epoxy Combination pH Electrode is Ideal for portable measurements as well as for routine measurements in-the-laboratory.
Brand:  WTW™ 103636
Product Code. 10557901
  • 305.00€ / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Combination pH electrode, gel filled with temperature compensation and 10-ft cable
  • For use with WTW ProfiLine and inoLab™ pH meter series
  • Ideal for portable measurements as well as for routine measurements in-the-laboratory
  • Includes gel electrolyte, DIN plug, banana plug, integrated temperature sensor SenTix™ 41-3, gel electrolyte, Built-in NTC (30kilohms) temperature probe, Fiber diaphragm, 3m fixed cable
  • Basic pH combination electrode type
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WTW ProfiLine and inoLab™ pH Meter Series
Ideal for portable measurements as well as for routine measurements in-the-laboratory
Gel
0 to 14
3 m
Basic pH Combination Electrode
Gel electrolyte, DIN plug, banana plug, integrated temperature sensor SenTix™ 41-3, gel electrolyte, Built-in NTC (30kilohms) temperature probe, Fiber diaphragm, 3m fixed cable
Epoxy
3 m