Learn More
Cole-Parmer™ Sensidyne™ 800885-111-2301 GilAir™ 5 Sampling Pump Kit
Ensure accuracy in high flow ranges with pumps featuring automated constant flow control. Cole-Parmer™ Sensidyne™ GilAir™ 5 Sampling Pump Kit includes pump, air boss (bag fill adapter), tubing, tool kit, rechargeable NiCd battery; 230VAC charger; and filter cassette holder.
Brand: Cole-Parmer™ 8627687
Description
- EMI/FRI protection prevents interference from plant operations
- See-through inlet filter housing protects pumps from dirt and debris while allowing easy inspection
- GilAir 5 is a double-diaphragm pump with a larger capacity battery to handle higher flow rates
- Clock features run and hold functions
Specifications
Pump, Air Boss (Bag Fill Adapter), Tubing, Tool Kit, Rechargeable NiCd Battery, 230 VAC Charger, Filter Cassette Holder | |
230 VAC | |
5.08 cm | |
0.75 to 3 L/min. (low-flow range: 1 to 500 mL/min.) | |
10.16 cm | |
9.32 cm | |
0 to +40°C |
UL (class I, groups A, B, C, D; class II, groups E, F, G; Class III, div 1); CENELEC | |
2 in. | |
Air Sampling Pump | |
4 in. | |
3043/64 in. | |
32° to 113°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.