Learn More
Brady™ Self-laminating Polyester Labels for M710
Printer Labels for use with Brady™ Printer M710.
Brand:  Brady™ 311976
 View more versions of this product
Product Code. 17817974
  • 128.00€ / Pack of 100
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Description
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Specifications
ASTM D 1000
Permanent Acrylic
White, Transparent
Label
Single-side Printable
Self-laminating Tag
Laboratory Labeling
1 Label
100 Labels
-321°F to +230°F
0.0019 in.
66.68 x 25.4 mm
1 in.
25.4 mm
1 in.
M7-115-461
None
Matte
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
-196°C
Rectangle
-196°C to +110°C
0.048 mm
2.625 in.
25.4 mm
25.4 mm
25.4 mm