Learn More
3M™ SecureFit™ 500 Foam Insert
Click to view available options
SF500-FOAM-EU
Description
- Insert fills the gap between the eyewear and the wearer’s face to increase protection levels
- Foam gasket provides improved mechanical impact absorption
- Foam gasket offers extra comfort and protection from sweat
- TPE gasket (with or without rx-prescription insert) available to protect against liquids and large dust particles
- Designed to fit specific ranges of 3M™ Safety Glasses
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Product Type | Accessory |
No. per Case | 20 |
Model | SF500-FOAM-EU |