missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SEA 0400

Potent Na+/Ca2+ exchanger (NCX) inhibitor.

258.00€ - 942.00€

Specifications

Product Type Inhibitor
Target Proteins Na+/Ca2+ Exchanger
CAS 223104-29-8
Molecular Formula C21H19F2NO3
Formula Weight 371.38g/mol
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18725806
View Documents Promotion Details
Tocris Bioscience™
6164/10
10 mg
258.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
18779016
View Documents
Tocris Bioscience™
6164/50
50 mg
942.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent Na+/Ca2+ exchanger (NCX) inhibitor. Inhibits NCX activity in rat astrocytes, microglia and cultured neurons, canine cardiac sarcolemmal vesicles and rat cardiomyocytes (IC50 values are 5 and 8.3, 33, 90 and 92 nM, respectively). Reduces infarct volumes and exhibits protective effects against myocardial ischemia in rats.
Specifications

Specifications

Inhibitor
223104-29-8
371.38g/mol
Na+/Ca2+ Exchanger Inhibitors
Na+/Ca2+ Exchanger
C21H19F2NO3
Store at -20°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only