missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDZ WAG 994, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Potent and selective A1 adenosine receptor agonist (Ki values are 23, > 10000 and 25000 nM for A1, A2A and A2B receptors respectively). Causes a sustained fall in blood pressure and heart rate in spontaneous hypertensive rats and inhibits adenosine deaminase-stimulated lipolysis in rat adipocytes (Ki = 8 nM).
Specifications
Specifications
CAS | 130714-47-5 |
Molecular Formula | C17H25N5O4 |
Quantity | 10 mg |
Synonym | sdz-wag-994, n-cyclohexyl-2'-o-methyladenosine, unii-567ewi9u1e, sdz wag 994, adenosine, n-cyclohexyl-2'-o-methyl, 2r,3r,4r,5r-5-6-cyclohexylamino purin-9-yl-2-hydroxymethyl-4-methoxyoxolan-3-ol, sdz wag-994, sdzwag994, n 6-cyclohexyl-2-o-methyladenosine, d08zbt |
InChI Key | JAKAFSGZUXCHLF-LSCFUAHRSA-N |
SMILES | COC1C(C(OC1N2C=NC3=C2N=CN=C3NC4CCCCC4)CO)O |
IUPAC Name | (2R,3R,4R,5R)-5-[6-(cyclohexylamino)purin-9-yl]-2-(hydroxymethyl)-4-methoxyoxolan-3-ol |
Molecular Weight (g/mol) | 363.418 |
PubChem CID | 164305 |
Formula Weight | 363.41 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'