missing translation for 'onlineSavingsMsg'
Learn More

SCX Antibody (PSH02-25), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-33137

Product Code. 30113999

  • 496.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SCX Monoclonal antibody specifically detects SCX in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCX
Monoclonal
1 mg/mL
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
BHLHA41, BHLHa48, Class A Basic Helix-Loop-Helix Protein 41, Class A Basic Helix-Loop-Helix Protein 48, Class II BHLH Protein Scleraxis, Scleraxis Basic Helix-Loop-Helix Transcription Factor, Scleraxis BHLH Transcription Factor, Scleraxis Homolog (Mouse), Scleraxis Homolog A, Scleraxis Homolog A (Mouse), Scleraxis Homolog B, Scleraxis Homolog B (Mouse), SCXA, SCXB
Recombinant protein within human SCX aa 1-201/ 201. (Uniprot: Q7RTU7)
100 μL
Primary
Human
Purified
Western Blot, Immunocytochemistry/Immunofluorescence
PSH02-25
Unconjugated
PBS (pH7.4), 0.1% BSA and 40% Glycerol
Rabbit
Protein A purified
RUO
642658
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.