Learn More
Lenz Laborglasintrumente Screwthread Tubes with NS Cone, Cap and Sealing
Click to view available options
Cone Size:
NS 19/26
NS 24/29
Unit Size:
Each
Description
- Screw thread adapters
- Made of DURAN™ tubing
- With screw-cap with aperture and silicone sealing ring
- With PTFE washers
Specifications
Specifications
| For Use With | For tubing dia. 5.5 to 6.5 mm |
| Packaging Quantity | 1 Ea. |
| Thread Size | 14 GL |
| Includes | NS cone, cap and sealing |
| Cone Size | NS 24/29 |