Learn More
Lenz Laborglasintrumente Screwthread Tubes For Glassblowers
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01300025
 View more versions of this product
Product Code. 10008420
  • 36.00€ / Pack of 10
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Description
  • Acc. to DIN 12216 and in addition to standard
  • With DIN thread (GL)
  • Material: DURAN™
Specifications
Specifications
22 mm
1.8 mm
25 GL
Borosilicate Glass (3.3)
10/Pk.