Learn More
BRAND™ Screw Cap Wash Head
Product Code. 10373073
Click to view available options
Thread Size (Metric):
GL 25
ETFE screw cap with FEP delivery tube for PFA wash bottles
  • GL 25
  • Spare wash head with delivery tube
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) Wash bottles of PFA
    Thread Size (Metric) GL 25