Learn More
3M™ Scotch-Brite™ Diamond Floor Brush
Scotch-Brite™ Diamond Floor Brush is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one step.
Brand: 3M™ 7100262371
1244.07 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Scotch-Brite™ Diamond Floor Brush is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one step.
- It is conformable, allowing the flexible bristles to reach low spots on the floor while minimizing debris clogging.
- No assembly or pre-soaking is required and it is easy to install on standard, low-speed equipment.
Specifications
Flexible Brush | |
Flexible | |
Scotch-Brite™ | |
Unscented | |
Yellow | |
3000 |
Bristle | |
No | |
Without Tip | |
Non-sterile | |
Low-Speed Machine | |
432 mm |