Learn More
SI Analytics™ ScienceLine Ion Selective Electrode
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
Brand:  SI Analytics™ 285130370
 View more versions of this product
Product Code. 10765493
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
  • Also measure pH and temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Potassium
K 60
0°C to 40°C
0.04 to 39,000 mg/L
2 to 12
Selective
Electrode