Learn More
SI Analytics™ ScienceLine Ion Selective Electrode
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
Brand:  SI Analytics™ 285130430
 View more versions of this product
Product Code. 10405253
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
  • Also measure pH and temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Copper
Cu 60
0°C to 80°C
0.0006 to 6400 mg/L
2 to 6
Selective
Electrode