missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SC 560, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Highly selective cyclooxygenase-1 (COX-1) inhibitor (IC50 values are 0.009 and 6.3 μM for COX-1 and COX-2 respectively). Inhibits COX-1-derived platelet thromboxane B2, gastric PGE2 and dermal PDE2 production. Significantly reduces ovarian surface epithelial tumor growth in vivo. Orally active.
Specifications
Specifications
CAS | 188817-13-2 |
Molecular Formula | C17H12ClF3N2O |
MDL Number | MFCD02179214 |
Quantity | 10 mg |
Synonym | 5-4-chlorophenyl-1-4-methoxyphenyl-3-trifluoromethyl-1h-pyrazole, razole, 5-4-chlorophenyl-1-4-methoxyphenyl-3-trifluoromethylpyrazole, chembl26915, 5-4-chlorophenyl-1-4-methoxyphenyl-3-trifluoromethyl pyrazole, lopac-s-2064, d0m5mi, sc hplc, 5-4-chlorophenyl-1-4-methoxyphenyl-3-trifluoromethyl-1h-py |
InChI Key | PQUGCKBLVKJMNT-UHFFFAOYSA-N |
SMILES | COC1=CC=C(C=C1)N1N=C(C=C1C1=CC=C(Cl)C=C1)C(F)(F)F |
IUPAC Name | 5-(4-chlorophenyl)-1-(4-methoxyphenyl)-3-(trifluoromethyl)-1H-pyrazole |
Molecular Weight (g/mol) | 352.74 |
PubChem CID | 4306515 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'