missing translation for 'onlineSavingsMsg'
Learn More

SC 19220, Tocris Bioscience™

Product Code. 18054782
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective EP1 receptor antagonist

Selective EP1 receptor antagonist (IC50 = 6.7 μM for inhibition of [3H]-PGE2 binding to EP1 transfected COS cells).

Chemical Identifiers

CAS 19395-87-0
Molecular Formula C16H14ClN3O3
Molecular Weight (g/mol) 331.756
InChI Key KNURFLJTOUGOOQ-UHFFFAOYSA-N
Synonym 2-acetylhydrazide 10 11h-carboxylic acid, 8-chloro-dibenz b,f 1,4 oxazepine-10 11h-carboxylic acid, tocris-1206, lopac-s-3065, d0oa8m, sc hplc , solid, n'-acetyl-3-chloro-6h-benzo b 1,4 benzoxazepine-5-carbohydrazide, n'-acetyl-3-chloro-6h-benzo b 1,5 benzoxazepine-5-carbohydrazide, 8-chlorodibenz b,f 1,4 oxazepine-10 11h-carboxy-2-acetyl hydrazide, 8-chloro-dibenzo z b,f 1,4 oxazepine-10 11h-carboxylic acid, 2-acetylhydrazide
PubChem CID 4336830
IUPAC Name N'-acetyl-3-chloro-6H-benzo[b][1,4]benzoxazepine-5-carbohydrazide
SMILES CC(=O)NNC(=O)N1CC2=CC=CC=C2OC3=C1C=C(C=C3)Cl

Specifications

Quantity 50 mg
Formula Weight 331.76
Percent Purity >99%
Chemical Name or Material SC 19220
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'