missing translation for 'onlineSavingsMsg'
Learn More

SB 269970 hydrochloride, Tocris Bioscience™

Product Code. 30001272
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective 5-HT7 antagonist; brain penetrant

Potent and selective 5-HT7 receptor antagonist (pKi values are 8.9, 7.2 and 6.0 for 5-HT7A, 5-HT5A and 5-HT1B and < 6.0 for 5-HT1A, 5-HT1D, 5-HT1E, 5-HT1F, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT4 and 5-HT6 receptors respectively). Brain penetrant in vivo.

Chemical Identifiers

CAS 261901-57-9
Molecular Formula C18H29ClN2O3S
Molecular Weight (g/mol) 388.951
InChI Key XQCJOYZLWFNDIO-PKLMIRHRSA-N
Synonym sb-269970 hydrochloride, sb 269970 hydrochloride, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-yl sulfonyl phenol hydrochloride, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidine-1-sulfonyl phenol hydrochloride, 3-2r-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-ylsulfonyl phenol hydrochloride, d0bg5h, sb-269970 hydrochloride, powder hplc, r-3-2-2-4-methylpiperidin-1-yl ethyl pyrrolidin-1-yl sulfonyl phenol hcl, r-3-2-2-4-methyl-piperidin-1-yl ethyl pyrrolidine-1-sulfonyl phenol hydrochloride
PubChem CID 11957684
IUPAC Name 3-[(2R)-2-[2-(4-methylpiperidin-1-yl)ethyl]pyrrolidin-1-yl]sulfonylphenol;hydrochloride
SMILES CC1CCN(CC1)CCC2CCCN2S(=O)(=O)C3=CC=CC(=C3)O.Cl

Specifications

Quantity 50 mg
Formula Weight 388.95
Percent Purity >98%
Chemical Name or Material SB 269970 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'