missing translation for 'onlineSavingsMsg'
Learn More

SB 265610, Tocris Bioscience™

Product Code. 30001412
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent CXCR2 antagonist

Potent CXCR2 antagonist that inhibits CINC-1-mediated but not C5a-mediated Ca2+ mobilization (IC50 values are 3.4 and 6800 nM respectively). Inhibits CINC-induced chemotaxis and attenuates neutrophil accumulation in inflammatory lung injury in vivo.

Chemical Identifiers

CAS 211096-49-0
Molecular Formula C14H9BrN6O
Molecular Weight (g/mol) 357.171
InChI Key SEDUMQWZEOMXSO-UHFFFAOYSA-N
Synonym chembl38182, n-2-bromophenyl-n'-7-cyano-1h-benzotriazol-4-yl urea, 1-2-bromophenyl-3-4-cyano-1h-benzo d 1,2,3 triazol-7-yl urea, 1-2-bromophenyl-3-7-cyano-1h-1,2,3-benzotriazol-4-yl urea, d02jxg, sb hplc, 1-2-bromo-phenyl-3-7-cyano-3h-benzotriazol-4-yl-urea, urea, n-2-bromophenyl-n'-4-cyano-1h-benzotriazol-7-yl, 1-2-bromophenyl-3-7-cyano-2h-1,2,3-benzotriazol-4-yl urea, 1-2-bromophenyl-3-7-cyano-3h-1,2,3-benzotriazol-4-yl urea
PubChem CID 9841667
IUPAC Name 1-(2-bromophenyl)-3-(7-cyano-2H-benzotriazol-4-yl)urea
SMILES C1=CC=C(C(=C1)NC(=O)NC2=CC=C(C3=NNN=C23)C#N)Br

Specifications

Quantity 1 mg
Formula Weight 357.16
Percent Purity >98%
Chemical Name or Material SB 265610
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'