Learn More
Sartorius Premium GLP Lab Printer for Secura™ and Quintix™ Semi Micro Balances
For use with Secura and Quintix Semi Micro Balances.
Brand: Sartorius YDP30
1091.89 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
For use with Secura and Quintix Semi Micro Balances.
Specifications
Secura and Quintix Semi-Micro Balance | |
Premium GLP Lab Printer |
Promotions
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Product Content Correction
Product Title
Sartorius Premium GLP Lab Printer for Secura™ and Quintix™ Semi Micro Balances > Premium GLP Lab Printer