Learn More
Sartorius Printer Paper
GREENER_CHOICE
Product Code. 11338963 Shop All Sartorius Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
YDP06 Dot Matrix Printer
YDP20-0CE Printer
Length (English):
550 in.
1944 in.
Length (Metric):
1397 cm
4937.76 cm
missing translation for 'unitSize'
Each
Pack of 5
Product Code. 11338963
missing translation for 'mfr': Sartorius 6906920
Paper for various Sartorius printers.
Paper for use with balance printers.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) YDP06 Dot Matrix Printer
Length (English) 550 in.
Length (Metric) 1397 cm
Type Printer Paper