Learn More
Sarcosine tert-butyl ester hydrochloride, 97%, Thermo Scientific Chemicals
79.00€ - 286.00€
Chemical Identifiers
CAS | 5616-81-9 |
---|---|
Molecular Formula | C7H16ClNO2 |
Molecular Weight (g/mol) | 181.66 |
MDL Number | MFCD00080936 |
InChI Key | RNLQHMIDSCYLAK-UHFFFAOYSA-N |
Synonym | h-sar-otbu.hcl, tert-butyl 2-methylamino acetate hydrochloride, sarcosine tert-butyl ester hydrochloride, tert-butyl sarcosinate hydrochloride, tert-butylsarcosinate hydrochloride, h-sar-otbuhcl, sarcosine t-butyl ester hydrochloride, n-methylglycine t-butyl ester hydrochloride, glycine, n-methyl-, 1,1-dimethylethyl ester, hydrochloride, h-sar-otbu hcl |
PubChem CID | 11252350 |
IUPAC Name | tert-butyl 2-(methylamino)acetate;hydrochloride |
SMILES | CC(C)(C)OC(=O)CNC.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15439728
|
Thermo Scientific Alfa Aesar
H63176.06 |
5 g |
79.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15449728
|
Thermo Scientific Alfa Aesar
H63176.14 |
25 g |
286.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sarcosine tert-butyl ester hydrochloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSarcosine tert-butyl ester hydrochloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol (0.3 gram in 2 mL MeOH).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
5616-81-9 | |
181.66 | |
RNLQHMIDSCYLAK-UHFFFAOYSA-N | |
11252350 | |
CC(C)(C)OC(=O)CNC.Cl |
C7H16ClNO2 | |
MFCD00080936 | |
h-sar-otbu.hcl, tert-butyl 2-methylamino acetate hydrochloride, sarcosine tert-butyl ester hydrochloride, tert-butyl sarcosinate hydrochloride, tert-butylsarcosinate hydrochloride, h-sar-otbuhcl, sarcosine t-butyl ester hydrochloride, n-methylglycine t-butyl ester hydrochloride, glycine, n-methyl-, 1,1-dimethylethyl ester, hydrochloride, h-sar-otbu hcl | |
tert-butyl 2-(methylamino)acetate;hydrochloride |
Specifications
5616-81-9 | |
White | |
CH3NHCH2CO2C(CH3)3·HCl | |
h-sar-otbu.hcl, tert-butyl 2-methylamino acetate hydrochloride, sarcosine tert-butyl ester hydrochloride, tert-butyl sarcosinate hydrochloride, tert-butylsarcosinate hydrochloride, h-sar-otbuhcl, sarcosine t-butyl ester hydrochloride, n-methylglycine t-butyl ester hydrochloride, glycine, n-methyl-, 1,1-dimethylethyl ester, hydrochloride, h-sar-otbu hcl | |
RNLQHMIDSCYLAK-UHFFFAOYSA-N | |
tert-butyl 2-(methylamino)acetate;hydrochloride | |
11252350 | |
97% | |
Sarcosine tert-butyl ester hydrochloride |
137°C to 141°C | |
C7H16ClNO2 | |
MFCD00080936 | |
Soluble in methanol (0.3 gram in 2 mL MeOH). | |
CC(C)(C)OC(=O)CNC.Cl | |
181.66 | |
181.66 | |
Powder |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only