Learn More
Thermo Scientific™ Sample and Skimmer Cones for iCAP™ Q/Qnova Series ICP-MS Systems
Perform robust, uncomplicated trace elemental analysis using high-quality consumables for the Thermo Scientific™ iCAP™ Q ICP-MS and Thermo Scientific™ iCAP™ Qnova series ICP-MS systems.
945.72€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15457904
|
Thermo Scientific™
1311870 |
Skimmer Cone, Nickel, Insert-Version |
945.72€
Each |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific™ iCAP™ Q/Qnova series ICP-MS systems. A range of easily changeable Ni and Pt Thermo Scientific™ sample and skimmer cones are available to suit your analysis requirements, all designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
- Sample cones made from nickel and platinum
- Skimmer cones made from nickel and platinum
- Platinum, boron free skimmer cone (for semiconductor-related applications)
- Skimmer cone inserts with different channel length to tailor instrument characteristics for each application
Specifications
iCAP Q/Qnova Series ICP-MS Systems |