Learn More
Thermo Scientific™ Sample and Reagent Cups for iCE™ 3000 Series AA systems
Ensure consistent sample presentation and complete sample pick-up with sample and reagent cups for Thermo Scientific™ iCE™ 3000 Series AA systems.
211.00€
Specifications
For Use With (Equipment) | GFS33 and GFS35(Z) Graphite Furnace Autosamplers |
---|
Product Code | Brand | Description | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Type | Price | Quantity & Availability | ||||
10461240
|
Thermo Scientific™
942339380031 |
Polypropylene Sample Cups for iCE™ 3000 Series AA Systems | Polypropylene Sample Cups |
211.00€
Pack of 1000 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
Description
Use Thermo Scientific™ Sample and Reagent Cups with Thermo Scientific™ GFS33 and GFS35(Z) Graphite Furnace Autosamplers to ensure consistent sample presentation and complete sample pick-up. Select from polypropylene sample and reagent cups, PTFE sample cups, and reduced volume sample cups.
The perfect choice for the Thermo Scientific™ GFS33 and GFS35(Z) Graphite Furnace Autosamplers, when used with our iCE 3000 Series Atomic Absorption spectrometers; these Thermo Scientific Sample and Reagent Cups for Thermo Scientific™ iCE™ 3000 Series AA systems ensure consistent sample presentation and complete sample pick-up. Select from polypropylene sample and reagent cups, PTFE sample cups, and reduced volume sample cups, according to your needs.Specifications
GFS33 and GFS35(Z) Graphite Furnace Autosamplers |