Learn More
Thermo Scientific™ Sample Loop Assemblies for Aqueous Samples
Designed specifically for aqueous samples, sample loop assemblies are available in a range of volumes to match your analysis requirements.
213.00€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
16471155
|
Thermo Scientific™
842347004052 |
ASXpress - Sample Loop 2mm ID/4.0ml |
213.00€
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Customize your consumables to match your chosen sample matrix precisely to maximize efficiency and precision. These Thermo Scientific™ Sample Loop Assemblies for Aqueous Samples are available in a range of volumes to match your analysis requirements.
Choose Customized Consumables to Suit your Analysis
Sample loop assembly aqueous 2mm ID, 0.7ml
Sample loop assembly aqueous 2mm ID, 1.0ml
Sample loop assembly aqueous 2mm ID, 1.5ml
Sample loop assembly aqueous 2mm ID, 2.0ml
Sample loop assembly aqueous 2mm ID, 3.0ml
Sample loop assembly aqueous 2mm ID, 4.0ml
Sample loop assembly aqueous 2mm ID, 5.0ml
Specifications
Aqueous Sample Loop | |
Mass Spectrometers |