missing translation for 'onlineSavingsMsg'
Learn More

Samarium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™

Product Code. 12927423
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Concentration 10mg/L
Packaging Bottle
Quantity 50 mL
Solvent or Matrix HNO3
Chemical Name or Material Samarium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'