Learn More
SymLine Safety Waste Collector
SymLine is the modular disposal system for liquid waste. It connects several laboratory workstations, HPLC systems and fume cupboards to a central disposal or collection point, such as a safety cabinet. Made of electrostatic conductive PTFE connects capillaries and tubes of the HPLC system with the disposal system.
Brand: SymLine 306494-EL
90.06 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
SymLine Safety Waste Collectors route the waste tubes from your HPLC system directly into the disposal system. A choice of several connections offers you complete freedom. Fittings for different capillary diameters are included in the scope of delivery.
- Made of electrically conductive PTFE-EL material to prevent static discharge
- Available with different numbers of tube connections
- For use on a SymLine pipe system for liquid waste disposal
Specifications
Black | |
PTFE-EL | |
Safety Waste Collector |
5 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm | |
GL45 |