Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
Brand:  SymLine™ 306494
90.06 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 16876011
  • 112.00€ / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Description
  • Available with different thread sizes for attachment to pipe system
  • Different numbers of hose connections for disposal of liquid waste
  • Available with electronic level sensors that indicate the degree of filling of the disposal container
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Specifications
Waste Disposal
5 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Collector for HPLC Waste
Safety Waste Cap
SymLine System
GL45