Learn More
SI Analytics™ S4D Stand Set
SI Analytics™ S4D Stand Set is designed for use with Lab and ProLab instruments and comes equipt with an arm and electrode holder.
Brand:  SI Analytics™ 285201520
Product Code. 10778794
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • For docking to the meters of the Lab and ProLab family
  • Not suitable for Lab 745, 845, 945 and ProLab 5000
  • Can be for autonomous usage as well
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S4D Stand Set
Arm and electrode holder