missing translation for 'onlineSavingsMsg'
Learn More

(S)-SNAP 5114, Tocris Bioscience™

Product Code. 18084882
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GABA uptake inhibitor

GABA transport inhibitor, showing selectivity for GAT-3 and GAT-2 (IC50 values are 5, 21 and 388 μM for hGAT-3, rGAT-2 and hGAT-1 respectively). Increases thalamic GABA levels and is an anticonvulsant following systemic administration in vivo.

Chemical Identifiers

CAS 157604-55-2
Molecular Formula C30H35NO6
Molecular Weight (g/mol) 505.61
MDL Number MFCD06411702
InChI Key VDLDUZLDZBVOAS-UHFFFAOYNA-N
Synonym s-snap-5114, chembl75035, s-snap 5114, s-1-2-tris 4-methoxyphenyl methoxy ethyl-3-piperidinecarboxylic acid, 3s-1-2-tris 4-methoxyphenyl methoxy ethyl piperidine-3-carboxylic acid, s-1-2-tris 4-methoxyphenyl methoxy ethyl piperidine-3-carboxylic acid, d0h7wk, s-snap hplc , solid, 1-2-tris 4-methoxyphenyl methoxy ethyl-s-3-piperidinecarboxylic acid, s-1-2-tris-4-methoxy-phenyl-methoxy-ethyl-piperidine-3-carboxylic acid
PubChem CID 10458835
IUPAC Name 1-{2-[tris(4-methoxyphenyl)methoxy]ethyl}piperidine-3-carboxylic acid
SMILES COC1=CC=C(C=C1)C(OCCN1CCCC(C1)C(O)=O)(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1

Specifications

Quantity 50 mg
Formula Weight 505.61
Percent Purity >98%
Chemical Name or Material (S)-SNAP 5114
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'