missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-N-Methyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine, Thermo Scientific™

Product Code. 15497525
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15497525

Brand: Thermo Scientific Alfa Aesar H25816.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 406681-25-2
Melting Point 114°C to 116°C
Molecular Formula C25H26FeNP
MDL Number MFCD09953458
Quantity 1 g
Synonym n-methyl-s-1-ferrocenyl-r-2-diphenylphosphino ethylamine
InChI Key NHBYRJAGLFUKCO-OLQNTQOENA-N
Optical Rotation +315° (c=0.75 in Ethanol)
SMILES [Fe].c1cccc1.CN[C@@H](C)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name cyclopentane;(1R)-1-cyclopentyl-2-diphenylphosphanyl-N-methylethanamine;iron
Molecular Weight (g/mol) 427.31
PubChem CID 90472614
Formula Weight 427.31
Chemical Name or Material (S)-(+)-N-Methyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.