missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-4-Fluorophenylglycine, 95%, (98% ee), Thermo Scientific Chemicals
208.00€ - 802.00€
Chemical Identifiers
CAS | 19883-57-9 |
---|---|
Molecular Formula | C8H8FNO2 |
Molecular Weight (g/mol) | 169.16 |
MDL Number | MFCD00213799 |
InChI Key | JKFYKCYQEWQPTM-ZETCQYMHSA-N |
Synonym | s-4-fluorophenylglycine, s-2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-l-alpha-phenylglycine, l-4-fluorophenylglycine, 4-fluoro-l-phenylglycine, l +-4-fluorophenylglycine, 2s-2-amino-2-4-fluorophenyl acetic acid, s-+-4-fluorophenyl glycine, p-fluorophenyl glycine, 4-fluorophenylglycine |
PubChem CID | 853015 |
IUPAC Name | (2S)-2-amino-2-(4-fluorophenyl)acetic acid |
SMILES | [NH3+][C@H](C([O-])=O)C1=CC=C(F)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10742643
|
Thermo Scientific Acros
362050010 |
1 g | Glass bottle |
208.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10621083
|
Thermo Scientific Acros
362050050 |
5 g | Glass bottle |
802.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19883-57-9 | |
169.16 | |
JKFYKCYQEWQPTM-ZETCQYMHSA-N | |
853015 | |
[NH3+][C@H](C([O-])=O)C1=CC=C(F)C=C1 |
C8H8FNO2 | |
MFCD00213799 | |
s-4-fluorophenylglycine, s-2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-l-alpha-phenylglycine, l-4-fluorophenylglycine, 4-fluoro-l-phenylglycine, l +-4-fluorophenylglycine, 2s-2-amino-2-4-fluorophenyl acetic acid, s-+-4-fluorophenyl glycine, p-fluorophenyl glycine, 4-fluorophenylglycine | |
(2S)-2-amino-2-(4-fluorophenyl)acetic acid |
Specifications
19883-57-9 | |
White | |
Authentic | |
MFCD00213799 | |
JKFYKCYQEWQPTM-ZETCQYMHSA-N | |
(2S)-2-amino-2-(4-fluorophenyl)acetic acid | |
853015 | |
95%,(98% ee) | |
(S)-4-Fluorophenylglycine, 95%, (98% e.e.) |
>300°C | |
97.5% min. (HPLC) | |
C8H8FNO2 | |
s-4-fluorophenylglycine, s-2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-l-alpha-phenylglycine, l-4-fluorophenylglycine, 4-fluoro-l-phenylglycine, l +-4-fluorophenylglycine, 2s-2-amino-2-4-fluorophenyl acetic acid, s-+-4-fluorophenyl glycine, p-fluorophenyl glycine, 4-fluorophenylglycine | |
[NH3+][C@H](C([O-])=O)C1=CC=C(F)C=C1 | |
169.16 | |
169.15 | |
Fluffy Powder |
Safety and Handling
HYGROSCOPIC
missing translation for 'tsca' : TSCA
RUO – Research Use Only