missing translation for 'onlineSavingsMsg'
Learn More
(S)-3-Amino-5-hexynoic acid hydrochloride, 95%, Thermo Scientific Chemicals Product Code.: 15458497

(S)-3-Amino-5-hexynoic acid hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15458497
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15458497

Brand: Thermo Scientific Alfa Aesar H52010.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

Solubility
Soluble in water (90 g/L at 25°C).

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270596-46-8
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.601
MDL Number MFCD01861006
InChI Key FVKOZZHCHSRKJA-JEDNCBNOSA-N
Synonym s-3-amino-5-hexynoic acid hydrochloride, s-3-aminohex-5-ynoic acid hydrochloride, 3s-3-aminohex-5-ynoic acid hydrochloride, s-3-amino-5-hexynoic acid-hcl, l-beta-homopropargylglycine hydrochloride, 5-hexynoic acid, 3-amino-, 3s-, hydrochloride 1:1, s-3-amino-5-hexynoic acidhcl, l-ss-homopropargylglycine hydrochlorideShow MoreShow Less
PubChem CID 2761700
IUPAC Name (3S)-3-aminohex-5-ynoic acid;hydrochloride
SMILES C#CCC(CC(=O)O)N.Cl

Specifications

Solubility Information Soluble in water (90g/L at 25°C).
Quantity 1 g
Formula Weight 163.6
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (S)-3-Amino-5-hexynoic acid hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberF15W036 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52010.03,H52010.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-3-Amino-5-hexynoic acid hydrochloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.