missing translation for 'onlineSavingsMsg'
Learn More

S 24795, Tocris Bioscience™

Product Code. 18093972
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Partial agonist at α7 nAChR

Partial agonist of α7 nAChR. Improves mnemonic function in aged mice for the treatment of aging-related memory disturbances. Displays more efficient memory-enhancing effects than memantine (Cat. No. 0773) in Alzheimer's disease mouse models.

Chemical Identifiers

CAS 304679-75-2
Molecular Formula C14H13BrINO
Molecular Weight (g/mol) 418.072
InChI Key LERUHBBUGAOYOD-UHFFFAOYSA-M
Synonym 2-2-4-bromophenyl-2-oxoethyl-1-methylpyridiniumiodide, 2-2-4-bromophenyl-2-oxoethyl-1-methylpyridinium iodide, 2-2-4-bromophenyl-2-oxoethyl-1-methylpyridin-1-ium iodide, s hplc
PubChem CID 9888248
IUPAC Name 1-(4-bromophenyl)-2-(1-methylpyridin-1-ium-2-yl)ethanone;iodide
SMILES C[N+]1=CC=CC=C1CC(=O)C2=CC=C(C=C2)Br.[I-]

Specifications

Quantity 10 mg
Formula Weight 418.07
Percent Purity >98%
Chemical Name or Material S 24795
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'