Learn More
Ruthenium, 2% on 3.18mm (0.125in) alumina pellets, Thermo Scientific Chemicals Product Code.: 11415350
Ruthenium, 2% on 3.18mm (0.125in) alumina pellets, Thermo Scientific Chemicals
Product Code. 11415350
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
Product Code. 11415350
Brand: Thermo Scientific Alfa Aesar 044575.14
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]
Specifications
Quantity 25 g
Odor Odorless
Chemical Name or Material Ruthenium, 2% on alumina pellets, 3.18mm (0.125 in.)
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU10I033 Certificate TypeCertificate of Analysis Date16/08/2022 Catalog Number044575.14,044575.36,044636.06,044636.03,044575.22
Lot NumberP14I046 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044575.14,044575.36,044636.06,044636.03,044575.22
Lot NumberW08H037 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044575.14,044575.36,044636.06,044636.03,044575.22
Lot NumberU20H052 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044575.14,044575.36,044636.06,044636.03,044575.22
Lot NumberW29H079 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044575.14,044575.36,044636.06,044636.03,044575.22
RUO – Research Use Only
Product Content Correction
Product Title
Ruthenium, 2% on 3.18mm (0.125in) alumina pellets, Thermo Scientific Chemicals >